Tisk…

5-Ethoxy-2-nitroaniline

Dodavatel: Apollo Scientific
Varování

OR110956-1G OR110956-5G
APOSOR110956-1GEA 622 CZK
APOSOR110956-1G APOSOR110956-5G
5-Ethoxy-2-nitroaniline
5-Ethoxy-2-nitroaniline
Vzorec: C₈H₁₀N₂O₃
MW: 182.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01830799
CAS číslo: 27076-16-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR