Tisk…

3-(Dimethylamino)-1-(2,4-dimethyl-1,3-thiazol-5-yl)prop-2-en-1-one

Dodavatel: Apollo Scientific

OR110949-1G OR110949-5G
APOSOR110949-1GEA 0 CZK
APOSOR110949-1G APOSOR110949-5G
3-(Dimethylamino)-1-(2,4-dimethyl-1,3-thiazol-5-yl)prop-2-en-1-one
3-(Dimethylamino)-1-(2,4-dimethyl-1,3-thiazol-5-yl)prop-2-en-1-one
Vzorec: C₁₀H₁₄N₂OS
MW: 210,296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179897
CAS číslo: 1432581-09-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR