Ethyl 8-chloroimidazo[1,2-a]pyridine-6-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR110907-1G
OR110907-100MG
OR110907-250MG
APOSOR110907-1GEA
8270
CZK
APOSOR110907-1G
APOSOR110907-100MG
APOSOR110907-250MG
Ethyl 8-chloroimidazo[1,2-a]pyridine-6-carboxylate
Ethyl 8-chloroimidazo[1,2-a]pyridine-6-carboxylate
Vzorec:
C₁₀H₉ClN₂O₂ MW: 224,644 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27995635 CAS číslo: 1823330-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...