4-Chloro-2-(1H-imidazol-1-yl)pyrimidine
Dodavatel: Apollo Scientific
OR110904-250MG
APOSOR110904-250MGEA
13460
CZK
APOSOR110904-250MG
4-Chloro-2-(1H-imidazol-1-yl)pyrimidine
4-Chloro-2-(1H-imidazol-1-yl)pyrimidine
Vzorec:
C₇H₅ClN₄ MW: 180,594 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09831925 CAS číslo: 114834-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...