Ethyl 5-bromo-6-hydroxynicotinate
Dodavatel: Apollo Scientific
|
Varování
|
OR110893-1G
OR110893-5G
OR110893-10G
APOSOR110893-1GEA
4570
CZK
APOSOR110893-1G
APOSOR110893-5G
APOSOR110893-10G
Ethyl 5-bromo-6-hydroxynicotinate
Ethyl 5-bromo-6-hydroxynicotinate
Vzorec:
C₈H₈BrNO₃ MW: 246,058 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02325626 CAS číslo: 169773-94-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...