Tisk…

5-Nitro-1-benzothiophene-2-carbohydrazide

Dodavatel: Apollo Scientific

OR110881-1G OR110881-5G OR110881-10G
APOSOR110881-1GEA 7050 CZK
APOSOR110881-1G APOSOR110881-5G APOSOR110881-10G
5-Nitro-1-benzothiophene-2-carbohydrazide
5-Nitro-1-benzothiophene-2-carbohydrazide
Vzorec: C₉H₇N₃O₃S
MW: 237,235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04085897
CAS číslo: 591761-80-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR