Tisk…

3-Methoxythiophene-2-carbonitrile

Dodavatel: Apollo Scientific
Nebezpečí

OR110878-1G OR110878-5G OR110878-250MG
APOSOR110878-1GEA 5790 CZK
APOSOR110878-1G APOSOR110878-5G APOSOR110878-250MG
3-Methoxythiophene-2-carbonitrile
3-Methoxythiophene-2-carbonitrile
Vzorec: C₆H₅NOS
MW: 139,175 g/mol
Bod varu: 72…74 °C (0,1 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD24052834
CAS číslo: 57059-18-4
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR