3-Bromo-4-(methoxymethoxy)benzaldehyde
Dodavatel: Apollo Scientific
OR110853-1G
OR110853-5G
APOSOR110853-1GEA
8830
CZK
APOSOR110853-1G
APOSOR110853-5G
3-Bromo-4-(methoxymethoxy)benzaldehyde
3-Bromo-4-(methoxymethoxy)benzaldehyde
Vzorec:
C₉H₉BrO₃ MW: 245,07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21195365 CAS číslo: 162269-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...