Methyl 5-bromo-2-hydroxy-3-(hydroxyamino)benzoate
Dodavatel: Apollo Scientific
|
Varování
|
OR110840-100MG
OR110840-250MG
APOSOR110840-100MGEA
622
CZK
APOSOR110840-100MG
APOSOR110840-250MG
Methyl 5-bromo-2-hydroxy-3-(hydroxyamino)benzoate
Methyl 5-bromo-2-hydroxy-3-(hydroxyamino)benzoate
Vzorec:
C₈H₈BrNO₄ MW: 262.057 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21602390 CAS číslo: 1427460-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...