Tisk…

2-Bromo-N,5-dimethylnicotinamide

Dodavatel: Apollo Scientific
Varování

OR110837-1G OR110837-250MG OR110837-5G
APOSOR110837-1GEA 14880 CZK
APOSOR110837-1G APOSOR110837-250MG APOSOR110837-5G
2-Bromo-N,5-dimethylnicotinamide
2-Bromo-N,5-dimethylnicotinamide
Vzorec: C₈H₉BrN₂O
MW: 229.074 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619606
CAS číslo: 1379302-91-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR