4-(3-Chlorophenyl)-2-methyl-2,4-dihydro-3H-1,2,4-triazol-3-one
Dodavatel: Apollo Scientific
OR110812-1G
OR110812-250MG
APOSOR110812-1GEA
12200
CZK
APOSOR110812-1G
APOSOR110812-250MG
4-(3-Chlorophenyl)-2-methyl-2,4-dihydro-3H-1,2,4-triazol-3-one
4-(3-Chlorophenyl)-2-methyl-2,4-dihydro-3H-1,2,4-triazol-3-one
Vzorec:
C₉H₈ClN₃O MW: 209.632 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18074541 CAS číslo: 1355171-14-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...