Tisk…

4-(4-Bromophenyl)-1-isopropyl-5-oxo-4,5-dihydro-1H-1,2,4-triazole-3-carboxylic acid

Dodavatel: Apollo Scientific

OR110811-1G OR110811-250MG
APOSOR110811-1GEA 12200 CZK
APOSOR110811-1G APOSOR110811-250MG
4-(4-Bromophenyl)-1-isopropyl-5-oxo-4,5-dihydro-1H-1,2,4-triazole-3-carboxylic acid
4-(4-Bromophenyl)-1-isopropyl-5-oxo-4,5-dihydro-1H-1,2,4-triazole-3-carboxylic acid
Vzorec: C₁₂H₁₂BrN₃O₃
MW: 326,146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18074539
CAS číslo: 1427460-86-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR