Tisk…

Ethyl 7-bromo-1H-indazole-5-carboxylate

Dodavatel: Apollo Scientific
Varování

OR110806-1G OR110806-250MG
APOSOR110806-1GEA 14880 CZK
APOSOR110806-1G APOSOR110806-250MG
Ethyl 7-bromo-1H-indazole-5-carboxylate
Ethyl 7-bromo-1H-indazole-5-carboxylate
Vzorec: C₁₀H₉BrN₂O₂
MW: 269,095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22370023
CAS číslo: 1355170-90-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR