Tisk…

4-Imidazo[2,1-b][1,3]thiazol-6-ylbenzonitrile

Dodavatel: Apollo Scientific

OR110782-1G OR110782-5G
APOSOR110782-1GEA 0 CZK
APOSOR110782-1G APOSOR110782-5G
4-Imidazo[2,1-b][1,3]thiazol-6-ylbenzonitrile
4-Imidazo[2,1-b][1,3]thiazol-6-ylbenzonitrile
Vzorec: C₁₂H₇N₃S
MW: 225,269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062346
CAS číslo: 118001-67-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR