5-Oxo-1-(thien-3-ylmethyl)pyrrolidine-3-carboxylic acid
Dodavatel: Apollo Scientific
OR110777-1G
OR110777-5G
APOSOR110777-1GEA
10550
CZK
APOSOR110777-1G
APOSOR110777-5G
5-Oxo-1-(thien-3-ylmethyl)pyrrolidine-3-carboxylic acid
5-Oxo-1-(thien-3-ylmethyl)pyrrolidine-3-carboxylic acid
Vzorec:
C₁₀H₁₁NO₃S Storage Temperature: Ambient |
MDL Number:
MFCD12406824 CAS číslo: 845546-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...