Tisk…

Ethyl thieno[3,2-b]thiophene-2-carboxylate

Dodavatel: Apollo Scientific

OR110773-1G OR110773-5G
APOSOR110773-1GEA 5460 CZK
APOSOR110773-1G APOSOR110773-5G
Ethyl thieno[3,2-b]thiophene-2-carboxylate
Ethyl thieno[3,2-b]thiophene-2-carboxylate
Vzorec: C₉H₈O₂S₂
MW: 212,289 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06803452
CAS číslo: 201004-08-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR