Tisk…

4-Chloro-6-(2,6-dimethylmorpholin-4-yl)pyrimidin-2-amine

Dodavatel: Apollo Scientific
Varování

OR110704-1G OR110704-250MG OR110704-5G
APOSOR110704-1GEA 8270 CZK
APOSOR110704-1G APOSOR110704-250MG APOSOR110704-5G
4-Chloro-6-(2,6-dimethylmorpholin-4-yl)pyrimidin-2-amine
4-Chloro-6-(2,6-dimethylmorpholin-4-yl)pyrimidin-2-amine
Vzorec: C₁₀H₁₅ClN₄O
MW: 242,705 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13533113
CAS číslo: 1197574-90-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR