Ethyl 3-amino-4-methoxybenzoate
Dodavatel: Apollo Scientific
|
Varování
|
OR110702-1G
OR110702-5G
APOSOR110702-1GEA
2280
CZK
APOSOR110702-1G
APOSOR110702-5G
Ethyl 3-amino-4-methoxybenzoate
Ethyl 3-amino-4-methoxybenzoate
Vzorec:
C₁₀H₁₃NO₃ MW: 195,215 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00017267 CAS číslo: 16357-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...