Tisk…

N-Ethyl-1,3,4-thiadiazole-2,5-diamine

Dodavatel: Apollo Scientific

OR110700-1G OR110700-250MG OR110700-5G
APOSOR110700-1GEA 5290 CZK
APOSOR110700-1G APOSOR110700-250MG APOSOR110700-5G
N-Ethyl-1,3,4-thiadiazole-2,5-diamine
N-Ethyl-1,3,4-thiadiazole-2,5-diamine
Vzorec: C₄H₈N₄S
MW: 144,198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18805199
CAS číslo: 79010-42-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR