1-Benzyl-4-(3-nitropyridin-2-yl)piperazine
Dodavatel: Apollo Scientific
OR110694-1G
OR110694-5G
OR110694-10G
APOSOR110694-1GEA
3910
CZK
APOSOR110694-1G
APOSOR110694-5G
APOSOR110694-10G
1-Benzyl-4-(3-nitropyridin-2-yl)piperazine
1-Benzyl-4-(3-nitropyridin-2-yl)piperazine
Vzorec:
C₁₆H₁₈N₄O₂ MW: 298,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02180918 CAS číslo: 499771-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...