3-[2,3-Di(benzyloxy)phenyl]propanenitrile
Dodavatel: Apollo Scientific
OR110688-10G
OR110688-1G
OR110688-5G
APOSOR110688-10GEA
9860
CZK
APOSOR110688-10G
APOSOR110688-1G
APOSOR110688-5G
3-[2,3-Di(benzyloxy)phenyl]propanenitrile
3-[2,3-Di(benzyloxy)phenyl]propanenitrile
Vzorec:
C₂₃H₂₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00100648 CAS číslo: 249278-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...