Tisk…

Diethyl 3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylate

Dodavatel: Apollo Scientific

OR110669-10G OR110669-1G OR110669-5G
APOSOR110669-10GEA 15150 CZK
APOSOR110669-10G APOSOR110669-1G APOSOR110669-5G
Diethyl 3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylate
Diethyl 3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylate
Vzorec: C₁₄H₁₆O₄S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00067988
CAS číslo: 152487-69-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR