Diethyl {[(4-bromophenyl)amino]methylene}malonate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Diethyl {[(4-bromophenyl)amino]methylidene}propanedioate
OR110666-1G
OR110666-5G
OR110666-10G
APOSOR110666-1GEA
4240
CZK
APOSOR110666-1G
APOSOR110666-5G
APOSOR110666-10G
Diethyl {[(4-bromophenyl)amino]methylene}malonate
Diethyl {[(4-bromophenyl)amino]methylene}malonate
Vzorec:
C₁₄H₁₆BrNO₄ Bod tání: 103…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173358 CAS číslo: 101937-44-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...