Tisk…
Methyl 4-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)-3-nitrobenzoate
Dodavatel: Apollo Scientific
OR110652-1G OR110652-5G
APOSOR110652-1GEA 7050 CZK
APOSOR110652-1G APOSOR110652-5G
Methyl 4-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)-3-nitrobenzoate
Methyl 4-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)-3-nitrobenzoate
Vzorec: C₁₅H₂₁N₃O₆
MW: 339.344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19442758
CAS číslo: 1198093-70-7