Tisk…

Benzyl 4-[(aminocarbonyl)amino]piperidine-1-carboxylate 95%

Dodavatel: Apollo Scientific
Varování

OR110649-1G OR110649-250MG
APOSOR110649-1GEA 13230 CZK
APOSOR110649-1G APOSOR110649-250MG
Benzyl 4-[(aminocarbonyl)amino]piperidine-1-carboxylate 95%
Benzyl 4-[(aminocarbonyl)amino]piperidine-1-carboxylate
Vzorec: C₁₄H₁₉N₃O₃
MW: 277,319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18804164
CAS číslo: 1427460-41-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR