Tisk…

Ethyl 5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate

Dodavatel: Apollo Scientific

OR110633-1G OR110633-10G OR110633-5G
APOSOR110633-1GEA 0 CZK
APOSOR110633-1G APOSOR110633-10G APOSOR110633-5G
Ethyl 5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate
Ethyl 5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate
Vzorec: C₁₀H₁₂O₂S
MW: 196,266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06662279
CAS číslo: 19282-44-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR