3-[4-(4-Bromo-1H-pyrazol-1-yl)phenyl]acrylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR110609-1G
OR110609-5G
APOSOR110609-1GEA
9860
CZK
APOSOR110609-1G
APOSOR110609-5G
3-[4-(4-Bromo-1H-pyrazol-1-yl)phenyl]acrylic acid
3-[4-(4-Bromo-1H-pyrazol-1-yl)phenyl]acrylic acid
Vzorec:
C₁₂H₉BrN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD12756236 CAS číslo: 1216366-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...