Tisk…

4-(4-Bromo-1H-pyrazol-1-yl)benzaldehyde

Dodavatel: Apollo Scientific

OR110602-1G OR110602-10G OR110602-5G
APOSOR110602-1GEA 0 CZK
APOSOR110602-1G APOSOR110602-10G APOSOR110602-5G
4-(4-Bromo-1H-pyrazol-1-yl)benzaldehyde
4-(4-Bromo-1H-pyrazol-1-yl)benzaldehyde
Vzorec: C₁₀H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD12756238
CAS číslo: 1174064-63-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR