1-(4-Acetylphenyl)-5-oxopyrrolidine-3-carboxylic acid
Dodavatel: Apollo Scientific
OR110574-1G
OR110574-5G
OR110574-10G
APOSOR110574-1GEA
6350
CZK
APOSOR110574-1G
APOSOR110574-5G
APOSOR110574-10G
1-(4-Acetylphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(4-Acetylphenyl)-5-oxopyrrolidine-3-carboxylic acid
Vzorec:
C₁₃H₁₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD01571218 CAS číslo: 100394-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...