Tisk…

6-Bromo-2H-chromene-3-carbonitrile

Dodavatel: Apollo Scientific

OR110564-1G OR110564-5G
APOSOR110564-1GEA 4240 CZK
APOSOR110564-1G APOSOR110564-5G
6-Bromo-2H-chromene-3-carbonitrile
6-Bromo-2H-chromene-3-carbonitrile
Vzorec: C₁₀H₆BrNO
MW: 236,065 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01662293
CAS číslo: 57543-68-7
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR