3-[(5-Bromopyridin-2-yl)oxy]benzaldehyde
Dodavatel: Apollo Scientific
OR110555-1G
OR110555-5G
APOSOR110555-1GEA
10550
CZK
APOSOR110555-1G
APOSOR110555-5G
3-[(5-Bromopyridin-2-yl)oxy]benzaldehyde
3-[(5-Bromopyridin-2-yl)oxy]benzaldehyde
Vzorec:
C₁₂H₈BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD17169531 CAS číslo: 1427460-53-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...