Tisk…

[2-(3,4-Dimethylphenoxy)phenyl]methanol

Dodavatel: Apollo Scientific

OR110505-1G OR110505-5G
APOSOR110505-1GEA 7050 CZK
APOSOR110505-1G APOSOR110505-5G
[2-(3,4-Dimethylphenoxy)phenyl]methanol
[2-(3,4-Dimethylphenoxy)phenyl]methanol
Vzorec: C₁₅H₁₆O₂
MW: 228,286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11190412
CAS číslo: 1039866-07-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR