Tisk…

[2-(4-tert-Butylphenoxy)phenyl]methanol

Dodavatel: Apollo Scientific

OR110503-1G OR110503-5G
APOSOR110503-1GEA 7050 CZK
APOSOR110503-1G APOSOR110503-5G
[2-(4-tert-Butylphenoxy)phenyl]methanol
[2-(4-tert-Butylphenoxy)phenyl]methanol
Vzorec: C₁₇H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD01814821
CAS číslo: 478032-35-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR