Methyl 5-bromo-2,4-dichlorobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
OR110486-1G
OR110486-250MG
APOSOR110486-1GEA
7870
CZK
APOSOR110486-1G
APOSOR110486-250MG
Methyl 5-bromo-2,4-dichlorobenzoate
Methyl 5-bromo-2,4-dichlorobenzoate
Vzorec:
C₈H₅BrCl₂O₂ Bod tání: 72…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD12032203 CAS číslo: 1305712-91-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...