Tisk…

(2-Bromo-4-methylphenoxy)acetonitrile

Dodavatel: Apollo Scientific

OR110465-1G OR110465-5G
APOSOR110465-1GEA 6350 CZK
APOSOR110465-1G APOSOR110465-5G
(2-Bromo-4-methylphenoxy)acetonitrile
(2-Bromo-4-methylphenoxy)acetonitrile
Vzorec: C₉H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD09906442
CAS číslo: 60758-92-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR