Tisk…

N-Ethyl-4-pentylbenzamide

Dodavatel: Apollo Scientific

OR110453-1G OR110453-250MG OR110453-5G
APOSOR110453-1GEA 5290 CZK
APOSOR110453-1G APOSOR110453-250MG APOSOR110453-5G
N-Ethyl-4-pentylbenzamide
N-Ethyl-4-pentylbenzamide
Vzorec: C₁₄H₂₁NO
MW: 219,323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03551084
CAS číslo: 401587-40-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR