Tisk…

4-Pentylbenzamide

Dodavatel: Apollo Scientific

OR110451-1G OR110451-250MG OR110451-5G
APOSOR110451-1GEA 5290 CZK
APOSOR110451-1G APOSOR110451-250MG APOSOR110451-5G
4-Pentylbenzamide
4-Pentylbenzamide
Vzorec: C₁₂H₁₇NO
MW: 191,269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00984217
CAS číslo: 138913-07-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR