Tisk…

2-Pyrrolidin-1-ylisonicotinamide

Dodavatel: Apollo Scientific

OR110450-1G OR110450-250MG
APOSOR110450-1GEA 5890 CZK
APOSOR110450-1G APOSOR110450-250MG
2-Pyrrolidin-1-ylisonicotinamide
2-Pyrrolidin-1-ylisonicotinamide
Vzorec: C₁₀H₁₃N₃O
MW: 191,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384920
CAS číslo: 1378457-21-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR