4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Dodavatel: Apollo Scientific
|
Varování
|
OR110421-1G
OR110421-5G
OR110421-10G
APOSOR110421-1GEA
3270
CZK
APOSOR110421-1G
APOSOR110421-5G
APOSOR110421-10G
4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Vzorec:
C₈H₆BrN₃O MW: 240,057 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18384918 CAS číslo: 1007578-99-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...