Tisk…

Quinoxaline-6-carboxamide

Dodavatel: Apollo Scientific
Varování

OR110372-1G OR110372-250MG
APOSOR110372-1GEA 4040 CZK
APOSOR110372-1G APOSOR110372-250MG
Quinoxaline-6-carboxamide
Quinoxaline-6-carboxamide
Vzorec: C₉H₇N₃O
MW: 173,171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09955631
CAS číslo: 457882-95-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR