2-Amino-5-chloronicotinohydrazide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Amino-5-chloropyridine-3-carbohydrazide
OR110366-250MG
APOSOR110366-250MGEA
1660
CZK
APOSOR110366-250MG
2-Amino-5-chloronicotinohydrazide
2-Amino-5-chloronicotinohydrazide
Vzorec:
C₆H₇ClN₄O MW: 186,599 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16631261 CAS číslo: 1314959-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...