Tisk…

2-Amino-5-bromo-4-methylnicotinonitrile

Dodavatel: Apollo Scientific
Varování

OR110318-1G OR110318-500MG
APOSOR110318-1GEA 10750 CZK
APOSOR110318-1G APOSOR110318-500MG
2-Amino-5-bromo-4-methylnicotinonitrile
2-Amino-5-bromo-4-methylnicotinonitrile
Vzorec: C₇H₆BrN₃
MW: 212,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09030795
CAS číslo: 180994-87-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR