Tisk…

2-(1,3-Benzoxazol-2-ylamino)ethanol

Dodavatel: Apollo Scientific
Varování

OR110298-1G OR110298-250MG
APOSOR110298-1GEA 16530 CZK
APOSOR110298-1G APOSOR110298-250MG
2-(1,3-Benzoxazol-2-ylamino)ethanol
2-(1,3-Benzoxazol-2-ylamino)ethanol
Vzorec: C₉H₁₀N₂O₂
MW: 178,188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12776538
CAS číslo: 134704-32-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR