Tisk…

4-(4-Methoxyphenyl)-1,3-thiazol-2(3H)-one

Dodavatel: Apollo Scientific

OR110297-1G OR110297-250MG
APOSOR110297-1GEA 8570 CZK
APOSOR110297-1G APOSOR110297-250MG
4-(4-Methoxyphenyl)-1,3-thiazol-2(3H)-one
4-(4-Methoxyphenyl)-1,3-thiazol-2(3H)-one
Vzorec: C₁₀H₉NO₂S
MW: 207,249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11632159
CAS číslo: 2104-03-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR