3-Acetamido-4-hydroxybenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-(5-Carboxy-2-hydroxyphenyl)acetamide, 2-(Acetylamino)-4-carboxyphenol, 3-(Acetylamino)-4-hydroxybenzoic acid
OR110291-1G
OR110291-250MG
APOSOR110291-1GEA
8070
CZK
APOSOR110291-1G
APOSOR110291-250MG
3-Acetamido-4-hydroxybenzoic acid
3-Acetamido-4-hydroxybenzoic acid
Vzorec:
C₉H₉NO₄ MW: 195.172 g/mol Bod tání: 238…241 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD14583147 CAS číslo: 91004-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...