Tisk…

4-Ethyl-N-methylbenzenesulphonamide

Dodavatel: Apollo Scientific

OR110290-1G OR110290-250MG OR110290-5G
APOSOR110290-1GEA 5590 CZK
APOSOR110290-1G APOSOR110290-250MG APOSOR110290-5G
4-Ethyl-N-methylbenzenesulphonamide
4-Ethyl-N-methylbenzenesulphonamide
Vzorec: C₉H₁₃NO₂S
MW: 199,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03550606
CAS číslo: 401583-57-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR