Tisk…

Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate

Dodavatel: Apollo Scientific
Varování

OR110272-1G OR110272-5G
APOSOR110272-1GEA 6620 CZK
APOSOR110272-1G APOSOR110272-5G
Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Vzorec: C₁₁H₁₁NO₃S
MW: 237.275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08706234
CAS číslo: 35212-88-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR