Tisk…

Methyl 4-amino-3-bromo-5-methylbenzoate

Dodavatel: Apollo Scientific
Varování

OR110270-10G OR110270-1G OR110270-5G
APOSOR110270-10GEA 2570 CZK
APOSOR110270-10G APOSOR110270-1G APOSOR110270-5G
Methyl 4-amino-3-bromo-5-methylbenzoate
Methyl 4-amino-3-bromo-5-methylbenzoate
Vzorec: C₉H₁₀BrNO₂
MW: 244,085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07021337
CAS číslo: 900019-52-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR