Ethyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR110260-1G
OR110260-5G
APOSOR110260-1GEA
1990
CZK
APOSOR110260-1G
APOSOR110260-5G
Ethyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
Ethyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
Vzorec:
C₉H₁₃NO₂ MW: 167,205 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00022376 CAS číslo: 2199-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...