Methyl 2-chloro-6-(piperazin-1-yl)isonicotinate
Dodavatel: Apollo Scientific
|
Varování
|
OR110254-1G
OR110254-250MG
APOSOR110254-1GEA
9100
CZK
APOSOR110254-1G
APOSOR110254-250MG
Methyl 2-chloro-6-(piperazin-1-yl)isonicotinate
Methyl 2-chloro-6-(piperazin-1-yl)isonicotinate
Vzorec:
C₁₁H₁₄ClN₃O₂ MW: 255,701 g/mol Bod tání: 72…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD12545990 CAS číslo: 1227954-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...